Thermo Scientific Chemicals

4-Fluorophthalic acid, 98%, Thermo Scientific Chemicals

Catalog number: H31597.03
1 g, Each
Thermo Scientific Chemicals

4-Fluorophthalic acid, 98%, Thermo Scientific Chemicals

Catalog number: H31597.03
1 g, Each
Quantity
Catalog number: H31597.03
also known as H31597-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
320-97-8
IUPAC Name
4-fluorobenzene-1,2-dicarboxylic acid
Molecular Formula
C8H5FO4
InChI Key
OMCXTFVBNCFZMY-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(F)C=C1C(O)=O
Assay (Silylated GC)
≥97.5%
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Form
Powder

Description

4-Fluorophthalic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorophthalic acid is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text