Thermo Scientific Chemicals

4-Fluoro-alpha-toluenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: H31605.MD
250 mg, Each
Thermo Scientific Chemicals

4-Fluoro-alpha-toluenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: H31605.MD
250 mg, Each
Quantity
Catalog number: H31605.MD
also known as H31605-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
103360-04-9
IUPAC Name
(4-fluorophenyl)methanesulfonyl chloride
Molecular Formula
C7H6ClFO2S
InChI Key
UUQGWVIRPCRTSA-UHFFFAOYSA-N
SMILES
FC1=CC=C(CS(Cl)(=O)=O)C=C1
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
53.0-63.0?C
Appearance (Color)
Yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text