Thermo Scientific Chemicals

4-Hydroxy-3-(trifluoromethyl)benzonitrile, 98+%, Thermo Scientific Chemicals

Catalog number: H31639.06
5 g, Each
Thermo Scientific Chemicals

4-Hydroxy-3-(trifluoromethyl)benzonitrile, 98+%, Thermo Scientific Chemicals

Catalog number: H31639.06
5 g, Each
Quantity
Catalog number: H31639.06
also known as H31639-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
124811-71-8
IUPAC Name
4-hydroxy-3-(trifluoromethyl)benzonitrile
Molecular Formula
C8H4F3NO
InChI Key
QXIFYONUKBXFTH-UHFFFAOYSA-N
SMILES
OC1=C(C=C(C=C1)C#N)C(F)(F)F
Melting Point (clear melt)
170-176?C
Appearance (Color)
Cream to brown or yellow
Assay (GC)
≥98.0%
Form
Powder

Description

As intermediate in organic syntheses. A novel electrolyte additive for lithium nickel manganese oxide cathode of high voltage lithium ion battery. It is oxidized preferably to carbonate solvents forming a low-impedance protective film.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
As intermediate in organic syntheses. A novel electrolyte additive for lithium nickel manganese oxide cathode of high voltage lithium ion battery. It is oxidized preferably to carbonate solvents forming a low-impedance protective film.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text