Thermo Scientific Chemicals

2,5-Dibromobenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: H31689.03
1 g, Each
Thermo Scientific Chemicals

2,5-Dibromobenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: H31689.03
1 g, Each
Quantity
Catalog number: H31689.03
also known as H31689-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
74553-29-0
IUPAC Name
2,5-dibromobenzaldehyde
Molecular Formula
C7H4Br2O
InChI Key
BQBXKWGMPUCSQV-UHFFFAOYSA-N
SMILES
BrC1=CC(C=O)=C(Br)C=C1
Appearance (Color)
White
Form
Powder
Melting Point (clear melt)
87.0-96.0?C
Assay (GC)
≥96.0%
Free acid (titration)
≤2.0%

Description

2,5-Dibromobenzaldehyde is used as intermediate, it is used to produce other chemicals, it can react with Ethane-1,2-diol to get 2-(2,5-Dibromo-phenyl)-[1,3]dioxolane. The reaction occurs with reagent p-toluenesulfonic acid, solvent toluene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dibromobenzaldehyde is used as intermediate, it is used to produce other chemicals, it can react with Ethane-1,2-diol to get 2-(2,5-Dibromo-phenyl)-[1,3]dioxolane. The reaction occurs with reagent p-toluenesulfonic acid, solvent toluene.

Solubility
Soluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text