2-Fluoro-4-hydroxypyridine, 97%, Thermo Scientific Chemicals
2-Fluoro-4-hydroxypyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Fluoro-4-hydroxypyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H31697.MD
also known as H31697-MD
Price (USD)/ Each
97.30
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
97.30
Add to cart
2-Fluoro-4-hydroxypyridine, 97%, Thermo Scientific Chemicals
Catalog numberH31697.MD
Price (USD)/ Each
97.30
-
Add to cart
Chemical Identifiers
CAS22282-69-5
IUPAC Name2-fluoro-1,4-dihydropyridin-4-one
Molecular FormulaC5H4FNO
InChI KeyBRLSDLHMGRDAPF-UHFFFAOYSA-N
SMILESFC1=CC(=O)C=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Melting Point (clear melt)143.0-152.0?C
Appearance (Color)White to pale cream
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only