Thermo Scientific Chemicals

2-Vinylbenzoic acid, 96%, Thermo Scientific Chemicals

Catalog number: H31723.03
1 g, Each
Thermo Scientific Chemicals

2-Vinylbenzoic acid, 96%, Thermo Scientific Chemicals

Catalog number: H31723.03
1 g, Each
Quantity
Catalog number: H31723.03
also known as H31723-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
27326-43-8
Form
Powder or lumpy powder
Assay (Silylated GC)
≥95.0%
Assay (Aqueous acid-base Titration)
≥95.0 to ≤105.0%
Appearance (Color)
White to pale yellow

Description

As intermediate in orgaic syntheses. Phthalides were obtained in low yields by palladium-assisted cyclization of 2-vinylbenzoic acid and 2-(2-methyl-2-propenyl)- benzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
As intermediate in orgaic syntheses. Phthalides were obtained in low yields by palladium-assisted cyclization of 2-vinylbenzoic acid and 2-(2-methyl-2-propenyl)- benzoic acid.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text