Thermo Scientific Chemicals

DL-beta-Homoproline hydrochloride, 97%

Catalog number: H31749.03
1 g, Each
Thermo Scientific Chemicals

DL-beta-Homoproline hydrochloride, 97%

Catalog number: H31749.03
1 g, Each
Quantity
Catalog number: H31749.03
also known as H31749-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
71985-79-0
IUPAC Name
2-(pyrrolidin-2-yl)acetic acid hydrochloride
Molecular Formula
C6H12ClNO2
InChI Key
VQDACVOAOJQTPR-UHFFFAOYNA-N
SMILES
Cl.OC(=O)CC1CCCN1
Form
Powder
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%
Comment
White
Melting Point (clear melt)
169.5-178.5?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text