Thermo Scientific Chemicals

5-Bromo-2-(trifluoromethoxy)benzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: H31751.09
10 g, Each
Thermo Scientific Chemicals

5-Bromo-2-(trifluoromethoxy)benzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: H31751.09
10 g, Each
Quantity
Catalog number: H31751.09
also known as H31751-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
923281-52-1
IUPAC Name
5-bromo-2-(trifluoromethoxy)benzaldehyde
Molecular Formula
C8H4BrF3O2
InChI Key
IBEWLWBZTHFCPI-UHFFFAOYSA-N
SMILES
FC(F)(F)OC1=C(C=O)C=C(Br)C=C1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥97.5%
Free acid (titration)
≤1%
Refractive Index
1.4960-1.5000 @ 20?C

Description

5-Bromo-2-(trifluoromethoxy)benzaldehyde is used in the synthesis of (-)-epicatechin, synthesis of biologically active compounds in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-(trifluoromethoxy)benzaldehyde is used in the synthesis of (-)-epicatechin, synthesis of biologically active compounds in vitro.

Solubility
Soluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text