Pyridine-3,4-dicarboximide, Thermo Scientific Chemicals
Pyridine-3,4-dicarboximide, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pyridine-3,4-dicarboximide, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number H31758.06
also known as H31758-06
Price (USD)/ Each
80.90
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
80.90
Add to cart
Pyridine-3,4-dicarboximide, Thermo Scientific Chemicals
Catalog numberH31758.06
Price (USD)/ Each
80.90
-
Add to cart
Chemical Identifiers
CAS4664-01-1
IUPAC Name1H,2H,3H-pyrrolo[3,4-c]pyridine-1,3-dione
Molecular FormulaC7H4N2O2
InChI KeySJSABZBUTDSWMJ-UHFFFAOYSA-N
SMILESO=C1NC(=O)C2=CN=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)227.0-236.0?C
FormCrystals or powder or crystalline powder
Assay (HPLC)≥96.0%
Appearance (Color)White to brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only