Ethyl 2-aminothiazole-5-carboxylate, 97%, Thermo Scientific Chemicals
Ethyl 2-aminothiazole-5-carboxylate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 2-aminothiazole-5-carboxylate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H31769.03
also known as H31769-03
Price (USD)/ Each
88.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
88.00
Add to cart
Ethyl 2-aminothiazole-5-carboxylate, 97%, Thermo Scientific Chemicals
Catalog numberH31769.03
Price (USD)/ Each
88.00
-
Add to cart
Chemical Identifiers
CAS32955-21-8
IUPAC Nameethyl 2-amino-1,3-thiazole-5-carboxylate
Molecular FormulaC6H8N2O2S
InChI KeyVNZXERIGKZNEKB-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CN=C(N)S1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)Yellow
Melting Point (clear melt)157.0-166.0?C
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only