In organic synthesis. Employed as important intermediates in various areas such as peptide synthesis, asymmetric synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
In organic synthesis. Employed as important intermediates in various areas such as peptide synthesis, asymmetric synthesis.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
RUO – Research Use Only
Clay M. Sharts,; William A. Sheppard. Modern Methods to Prepare Monofluoroaliphatic Compounds. Organic Reactions. 2011,