6-Bromopyridine-2-carboxamide, 97%, Thermo Scientific Chemicals
6-Bromopyridine-2-carboxamide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Bromopyridine-2-carboxamide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H31811.06
also known as H31811-06
Price (USD)/ Each
202.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
202.00
Add to cart
6-Bromopyridine-2-carboxamide, 97%, Thermo Scientific Chemicals
Catalog numberH31811.06
Price (USD)/ Each
202.00
-
Add to cart
Chemical Identifiers
CAS25194-52-9
IUPAC Name6-bromopyridine-2-carboxamide
Molecular FormulaC6H5BrN2O
InChI KeyHWRUOHMDYQEWOD-UHFFFAOYSA-N
SMILESNC(=O)C1=NC(Br)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)132.0-142.0?C
Appearance (Color)White
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only