Thermo Scientific Chemicals

Titanium(IV) chloride, 1M soln. in dichloromethane, Thermo Scientific Chemicals

Catalog number: H31830.AP
500 mL, Each
Thermo Scientific Chemicals

Titanium(IV) chloride, 1M soln. in dichloromethane, Thermo Scientific Chemicals

Catalog number: H31830.AP
500 mL, Each
Quantity
Catalog number: H31830.AP
also known as H31830-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7550-45-0
IUPAC Name
titanium(4+) tetrachloride
Molecular Formula
Cl4Ti
InChI Key
XJDNKRIXUMDJCW-UHFFFAOYSA-J
SMILES
[Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]
Appearance (Color)
Colorless to pale yellow to dark brown
Form
Liquid
Assay (Titration ex Chloride)
0.95-1.10M

Description

Activates pyrrolidines for improved conversion, via a modified Bouveault reaction, to the corresponding α,α-dimethylamines

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Activates pyrrolidines for improved conversion, via a modified Bouveault reaction, to the corresponding α,α-dimethylamines

Solubility
Reacts with water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text