Thermo Scientific Chemicals

Tris(2-dimethylaminoethyl)amine, 98+%, Thermo Scientific Chemicals

Catalog number: H31853.14
25 g, Each
Thermo Scientific Chemicals

Tris(2-dimethylaminoethyl)amine, 98+%, Thermo Scientific Chemicals

Catalog number: H31853.14
25 g, Each
Quantity
Catalog number: H31853.14
also known as H31853-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
33527-91-2
IUPAC Name
(2-{bis[2-(dimethylamino)ethyl]amino}ethyl)dimethylamine
Molecular Formula
C12H30N4
InChI Key
VMGSQCIDWAUGLQ-UHFFFAOYSA-N
SMILES
CN(C)CCN(CCN(C)C)CCN(C)C
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Refractive Index
1.4525-1.4555 @ 20?C

Description

Tris(2-dimethylaminoethyl)amine is used as an atom transfer radical polymerization (ATRP) ligand for the creation of telechelic polymers. It is a tripodal polyamine ligand and forms complexes with group 1 metals, which is used in alkali-metal-mediated synthetic applications. Further, it is involved in the macrocyclization processes by reacting with tripodal esters in methanol to give tricyclic cryptands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(2-dimethylaminoethyl)amine is used as an atom transfer radical polymerization (ATRP) ligand for the creation of telechelic polymers. It is a tripodal polyamine ligand and forms complexes with group 1 metals, which is used in alkali-metal-mediated synthetic applications. Further, it is involved in the macrocyclization processes by reacting with tripodal esters in methanol to give tricyclic cryptands.

Solubility
Miscible with water.

Notes
Hygroscopic. Air sensitive. Incompatible with oxidizing agents, acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text