Methyl 1,2,3,4-tetra-O-acetyl-beta-D-glucuronate, 98%, Thermo Scientific Chemicals
Methyl 1,2,3,4-tetra-O-acetyl-beta-D-glucuronate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 1,2,3,4-tetra-O-acetyl-beta-D-glucuronate, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Catalog number H31888.03
also known as H31888-03
Price (USD)/ Each
195.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
195.00
Add to cart
Methyl 1,2,3,4-tetra-O-acetyl-beta-D-glucuronate, 98%, Thermo Scientific Chemicals
Catalog numberH31888.03
Price (USD)/ Each
195.00
-
Add to cart
Chemical Identifiers
CAS7355-18-2
IUPAC Namemethyl 3,4,5,6-tetrakis(acetyloxy)oxane-2-carboxylate
Molecular FormulaC15H20O11
InChI KeyDPOQCELSZBSZGX-UHFFFAOYNA-N
SMILESCOC(=O)C1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Optical Rotation+8.5? ± 0.5? (c=1 in CHCl₃)
Assay (GC)≥97.5%
Appearance (Color)White
Melting Point (clear melt)175.5-181.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only