Thermo Scientific Chemicals

1-(2-Fluorophenyl)ethanol, 95%, Thermo Scientific Chemicals

Catalog number: H31900.09
10 g, Each
Thermo Scientific Chemicals

1-(2-Fluorophenyl)ethanol, 95%, Thermo Scientific Chemicals

Catalog number: H31900.09
10 g, Each
Quantity
Catalog number: H31900.09
also known as H31900-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
445-26-1
IUPAC Name
1-(2-fluorophenyl)ethan-1-ol
Molecular Formula
C8H9FO
InChI Key
SXFYVXSOEBCFLV-UHFFFAOYNA-N
SMILES
CC(O)C1=CC=CC=C1F
Assay (GC)
≥94.0%
Form
Liquid
Appearance (Color)
Clear colorless to yellow or orange
Refractive Index
1.5000-1.5060 @ 20?C

Description

It is an important raw material in organic synthesis, agricultural fields and in dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material in organic synthesis, agricultural fields and in dyestuffs.

Solubility
Immiscible in water.

Notes
Store in cool dry place. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text