Thermo Scientific Chemicals

4-Methyl-2-(trifluoromethyl)benzonitrile, 98%, Thermo Scientific Chemicals

Catalog number: H31904.06
5 g, Each
Thermo Scientific Chemicals

4-Methyl-2-(trifluoromethyl)benzonitrile, 98%, Thermo Scientific Chemicals

Catalog number: H31904.06
5 g, Each
Quantity
Catalog number: H31904.06
also known as H31904-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
261952-05-0
Form
Powder or fused solid
Melting Point (clear melt)
23.0-30.0?C
Appearance (Color)
White to pale cream
Assay (GC)
≥97.5%

Description

It is used as primary and secondary reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as primary and secondary reagent.

Solubility
Insoluble in water.

Notes
Store in cool dry place. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text