2-Amino-3-fluoropyridine, 97%, Thermo Scientific Chemicals
2-Amino-3-fluoropyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-3-fluoropyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H31970.06
also known as H31970-06
Price (USD)/ Each
187.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
187.00
Add to cart
2-Amino-3-fluoropyridine, 97%, Thermo Scientific Chemicals
Catalog numberH31970.06
Price (USD)/ Each
187.00
-
Add to cart
Chemical Identifiers
CAS21717-95-3
IUPAC Name3-fluoropyridin-2-amine
Molecular FormulaC5H5FN2
InChI KeyWWEINXQNCAWBPD-UHFFFAOYSA-N
SMILESNC1=C(F)C=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Melting Point (clear melt)41.5-47.5?C
FormCrystals or powder or crystalline powder
Appearance (Color)White to yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only