Thermo Scientific Chemicals

2-Bromo-3-fluoro-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals

Catalog number: H31993.03
1 g, Each
Thermo Scientific Chemicals

2-Bromo-3-fluoro-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals

Catalog number: H31993.03
1 g, Each
Quantity
Catalog number: H31993.03
also known as H31993-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
89402-29-9
IUPAC Name
2-bromo-3-fluoro-5-(trifluoromethyl)pyridine
Molecular Formula
C6H2BrF4N
InChI Key
DNVIECHQUYQOOH-UHFFFAOYSA-N
SMILES
FC1=C(Br)N=CC(=C1)C(F)(F)F
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless to yellow
Refractive Index
1.4615-1.4665 @ 20?C
Form
Liquid
Identification (FTIR)
Conforms

Description

2-Bromo-3-fluoro-5-(trifluoromethyl)pyridine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-3-fluoro-5-(trifluoromethyl)pyridine is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text