Thermo Scientific Chemicals

6-Amino-1-tetralone, 97%, Thermo Scientific Chemicals

Catalog number: H32028.09
10 g, Each
Thermo Scientific Chemicals

6-Amino-1-tetralone, 97%, Thermo Scientific Chemicals

Catalog number: H32028.09
10 g, Each
Quantity
Catalog number: H32028.09
also known as H32028-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3470-53-9
IUPAC Name
6-amino-1,2,3,4-tetrahydronaphthalen-1-one
Molecular Formula
C10H11NO
InChI Key
BEVVUJBVEXJGKM-UHFFFAOYSA-N
SMILES
NC1=CC=C2C(=O)CCCC2=C1
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
125.0-135.0?C
Appearance (Color)
Cream to yellow
Identification (FTIR)
Conforms
Assay (GC)
≥96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text