Thermo Scientific Chemicals

2-Bromo-2'-(trifluoromethyl)acetophenone, 97%, Thermo Scientific Chemicals

Catalog number: H32032.06
5 g, Each
Thermo Scientific Chemicals

2-Bromo-2'-(trifluoromethyl)acetophenone, 97%, Thermo Scientific Chemicals

Catalog number: H32032.06
5 g, Each
Quantity
Catalog number: H32032.06
also known as H32032-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
54109-16-9
IUPAC Name
2-bromo-1-[2-(trifluoromethyl)phenyl]ethan-1-one
Molecular Formula
C9H6BrF3O
InChI Key
KWZCBMKXNYOQAK-UHFFFAOYSA-N
SMILES
FC(F)(F)C1=CC=CC=C1C(=O)CBr
Assay (GC)
≥96.0%
Form
Fused solid
Melting Point (clear melt)
25.5-34.5?C
Appearance (Color)
White to yellow

Description

The sulfonyl urea thus obtained was used directly in the following condensation with 4-(trifluoromethyl)phenacyl bromide. Upon heating, benzenesulfinic acid ammonium salt might react with 2-(trifluoromethyl)phenacyl bromide to give β-keto sulfone. In Rap-Stoermer condensation reation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The sulfonyl urea thus obtained was used directly in the following condensation with 4-(trifluoromethyl)phenacyl bromide. Upon heating, benzenesulfinic acid ammonium salt might react with 2-(trifluoromethyl)phenacyl bromide to give β-keto sulfone. In Rap-Stoermer condensation reation.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text