Thermo Scientific Chemicals

4-Bromo-2-hydroxypyridine, 97%, Thermo Scientific Chemicals

Catalog number: H32067.03
1 g, Each
Thermo Scientific Chemicals

4-Bromo-2-hydroxypyridine, 97%, Thermo Scientific Chemicals

Catalog number: H32067.03
1 g, Each
Quantity
Catalog number: H32067.03
also known as H32067-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
36953-37-4
IUPAC Name
4-bromo-1,2-dihydropyridin-2-one
Molecular Formula
C5H4BrNO
InChI Key
SSLMGOKTIUIZLY-UHFFFAOYSA-N
SMILES
BrC1=CC(=O)NC=C1
Assay (Non-aqueous acid-base Titration)
>96.0 to <104.0%
Form
Powder
Appearance (Color)
Brown
Assay (Silylated GC)
>96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text