Thermo Scientific Chemicals

2,2,2-Trimethylthioacetamide, 97%, Thermo Scientific Chemicals

Catalog number: H32092.09
10 g, Each
Thermo Scientific Chemicals

2,2,2-Trimethylthioacetamide, 97%, Thermo Scientific Chemicals

Catalog number: H32092.09
10 g, Each
Quantity
Catalog number: H32092.09
also known as H32092-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
630-22-8
IUPAC Name
2,2-dimethylpropanethioamide
Molecular Formula
C5H11NS
InChI Key
FJZJUSOFGBXHCV-UHFFFAOYSA-N
SMILES
CC(C)(C)C(N)=S
Form
Crystals or powder or crystalline powder
Assay (GC)
≥96.0%
Melting Point (clear melt)
112.0-121.0?C
Appearance (Color)
White

Description

2,2,2-Trimethylthioacetamide, is used as a pharmaceutical and organic intermediate, an important amino protective agent, pharmaceutical and synthetic organic make intermediates, organic synthesis used to introduce t- Boc protect gene species.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,2-Trimethylthioacetamide, is used as a pharmaceutical and organic intermediate, an important amino protective agent, pharmaceutical and synthetic organic make intermediates, organic synthesis used to introduce t- Boc protect gene species.

Solubility
Soluble in water (10 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text