4-Fluorothiobenzamide, 97%, Thermo Scientific Chemicals
4-Fluorothiobenzamide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Fluorothiobenzamide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H32093.03
also known as H32093-03
Price (USD)/ Each
38.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
38.00
Add to cart
4-Fluorothiobenzamide, 97%, Thermo Scientific Chemicals
Catalog numberH32093.03
Price (USD)/ Each
38.00
-
Add to cart
Chemical Identifiers
CAS22179-72-2
IUPAC Name4-fluorobenzene-1-carbothioamide
Molecular FormulaC7H6FNS
InChI KeyVQFOHZWOKJQOGO-UHFFFAOYSA-N
SMILESNC(=S)C1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)145.0-151.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only