Thermo Scientific Chemicals

Stains-All, 96%

Catalog number: H32127.03
1g, Each
Thermo Scientific Chemicals

Stains-All, 96%

Catalog number: H32127.03
1g, Each
Quantity
Catalog number: H32127.03
also known as H32127-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7423-31-6
IUPAC Name
1-ethyl-2-{3-[(2Z)-1-ethyl-1H,2H-naphtho[1,2-d][1,3]thiazol-2-ylidene]-2-methylprop-1-en-1-yl}naphtho[1,2-d][1,3]thiazol-1-ium bromide
Molecular Formula
C30H27BrN2S2
InChI Key
MPBRYMWMMKKRGC-UHFFFAOYSA-M
SMILES
[Br-].CCN1\C(SC2=CC=C3C=CC=CC3=C12)=C\C(C)=CC1=[N+](CC)C2=C(S1)C=CC1=CC=CC=C21
Form
Powder
Elemental Analysis
Nitrogen, N: 4.62-5.11% (Theory: 4.86%)
Appearance (Color)
Dark blue or dark grey to black
Elemental Analysis
Carbon, C: 61.17-67.61% (Theory: 64.39%)
Assay (Dye)
≥95.0%

Description

General protein and nucleic acid stain, which stains different colors on bands of RNA (bluish purple), DNA (blue) and proteins (red)

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text