Thermo Scientific Chemicals

1,2,3,4-Tetrahydro-1-naphthoic acid, 98%, Thermo Scientific Chemicals

Catalog number: H32138.MD
250 mg, Each
Thermo Scientific Chemicals

1,2,3,4-Tetrahydro-1-naphthoic acid, 98%, Thermo Scientific Chemicals

Catalog number: H32138.MD
250 mg, Each
Quantity
Catalog number: H32138.MD
also known as H32138-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1914-65-4
IUPAC Name
1,2,3,4-tetrahydronaphthalene-1-carboxylic acid
Molecular Formula
C11H12O2
InChI Key
VDLWTJCSPSUGOA-UHFFFAOYNA-N
SMILES
OC(=O)C1CCCC2=CC=CC=C12
Appearance (Color)
Pale cream
Melting Point (clear melt)
79.0-86.0?C
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Form
Powder
Assay (Silylated GC)
≥97.5%

Description

1,2,3,4-Tetrahydro-1-naphthoic acid is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. 1,2,3,4-tetrahydronaphthalene-1-carboxylic acid is a reagent used to produce protease inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3,4-Tetrahydro-1-naphthoic acid is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. 1,2,3,4-tetrahydronaphthalene-1-carboxylic acid is a reagent used to produce protease inhibitors.

Solubility
Soluble in methanol, and chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text