Thermo Scientific Chemicals

11-Chloro-1,1'-di-n-propyl-3,3,3',3'-tetramethyl-10,12-trimethyleneindatricarbocyanine iodide, 95%, Thermo Scientific Chemicals

Catalog number: H32150.06
5 g, Each
Thermo Scientific Chemicals

11-Chloro-1,1'-di-n-propyl-3,3,3',3'-tetramethyl-10,12-trimethyleneindatricarbocyanine iodide, 95%, Thermo Scientific Chemicals

Catalog number: H32150.06
5 g, Each
Quantity
Catalog number: H32150.06
also known as H32150-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
207399-07-3
IUPAC Name
2-(2-{2-chloro-3-[2-(3,3-dimethyl-1-propyl-2,3-dihydro-1H-indol-2-ylidene)ethylidene]cyclohex-1-en-1-yl}ethenyl)-3,3-dimethyl-1-propyl-3H-indol-1-ium iodide
Molecular Formula
C36H44ClIN2
InChI Key
IRPKBYJYVJOQHQ-UHFFFAOYSA-M
SMILES
[I-].CCCN1C2=CC=CC=C2C(C)(C)C1=CC=C1CCCC(C=CC2=[N+](CCC)C3=CC=CC=C3C2(C)C)=C1Cl
Elemental Analysis
(Iodine) : 18.52-19.52%
Form
Crystals or crystalline powder
Chloride content
4.81-5.81%
Elemental Analysis
(Nitrogen) : 3.70-4.70%
Appearance (Color)
Green/gold to dark green

Description

11-Chloro-1,1'-di-n-propyl-3,3,3',3'-tetramethyl-10,12-trimethyleneindatricarbocyanine iodide is used as a laser and IR dye. It is also used as lithographic computer to plate laser applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
11-Chloro-1,1′-di-n-propyl-3,3,3′,3′-tetramethyl-10,12-trimethyleneindatricarbocyanine iodide is used as a laser and IR dye. It is also used as lithographic computer to plate laser applications.

Solubility
It is soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text