5-Bromo-2,4-difluorotoluene, 98%, Thermo Scientific Chemicals
5-Bromo-2,4-difluorotoluene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Bromo-2,4-difluorotoluene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H32155.06
also known as H32155-06
Price (USD)/ Each
78.80
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
78.80
Add to cart
5-Bromo-2,4-difluorotoluene, 98%, Thermo Scientific Chemicals
Catalog numberH32155.06
Price (USD)/ Each
78.80
-
Add to cart
Chemical Identifiers
CAS159277-47-1
IUPAC Name1-bromo-2,4-difluoro-5-methylbenzene
Molecular FormulaC7H5BrF2
InChI KeyWOCZKGUFVNUBEC-UHFFFAOYSA-N
SMILESCC1=CC(Br)=C(F)C=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.5060-1.5100 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only