3,5-Dimethylpyridine-4-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals
3,5-Dimethylpyridine-4-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,5-Dimethylpyridine-4-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
500 mg
Catalog number H32159.ME
also known as H32159-ME
Price (USD)/ Each
882.65
Online exclusive
980.00 
Save 97.35 (10%)
-
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (USD)/ Each
882.65
Online exclusive
980.00 
Save 97.35 (10%)
Add to cart
3,5-Dimethylpyridine-4-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals
Catalog numberH32159.ME
Price (USD)/ Each
882.65
Online exclusive
980.00 
Save 97.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1032358-02-3
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)121.0-133.0?C
Assay (GC)≥94.0%
Appearance (Color)White to cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only