3,4-Dihydro-2H-1,5-benzodioxepin-7-carboxaldehyde, 95%, Thermo Scientific Chemicals
3,4-Dihydro-2H-1,5-benzodioxepin-7-carboxaldehyde, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,4-Dihydro-2H-1,5-benzodioxepin-7-carboxaldehyde, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H32167.03
also known as H32167-03
Price (USD)/ Each
126.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
126.00
Add to cart
3,4-Dihydro-2H-1,5-benzodioxepin-7-carboxaldehyde, 95%, Thermo Scientific Chemicals
Catalog numberH32167.03
Price (USD)/ Each
126.00
-
Add to cart
Chemical Identifiers
CAS67869-90-3
IUPAC Name3,4-dihydro-2H-1,5-benzodioxepine-7-carbaldehyde
Molecular FormulaC10H10O3
InChI KeyLCSVYSVGXQQHSI-UHFFFAOYSA-N
SMILESO=CC1=CC2=C(OCCCO2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to brown
Assay (GC)≥94.0%
FormFused solid or clear liquid as melt
Refractive Index1.5780-1.5840 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only