Potassium bromomethyltrifluoroborate, 95%, Thermo Scientific Chemicals
Potassium bromomethyltrifluoroborate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium bromomethyltrifluoroborate, 95%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number H32168.06
also known as H32168-06
Price (USD)/ Each
249.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
249.00
Add to cart
Potassium bromomethyltrifluoroborate, 95%, Thermo Scientific Chemicals
Catalog numberH32168.06
Price (USD)/ Each
249.00
-
Add to cart
Chemical Identifiers
CAS888711-44-2
IUPAC Namepotassium (bromomethyl)trifluoroboranuide
Molecular FormulaCH2BBrF3K
InChI KeyAZDFPIRYUOCVCJ-UHFFFAOYSA-N
SMILES[K+].F[B-](F)(F)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay from Supplier's CofA≥94.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only