Thermo Scientific Chemicals

2,2-Difluoro-4-pentenoic acid, 97%, Thermo Scientific Chemicals

Catalog number: H32173.06
5 g, Each
Thermo Scientific Chemicals

2,2-Difluoro-4-pentenoic acid, 97%, Thermo Scientific Chemicals

Catalog number: H32173.06
5 g, Each
Quantity
Catalog number: H32173.06
also known as H32173-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
55039-89-9
IUPAC Name
2,2-difluoropent-4-enoic acid
Molecular Formula
C5H6F2O2
InChI Key
LHOKYUDUAYXFGF-UHFFFAOYSA-N
SMILES
OC(=O)C(F)(F)CC=C
Form
Liquid
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless to yellow
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Refractive Index
1.3845-1.3895 @ 20?C

Description

2,2-Difluoro-4-pentenoic acid is involved in the iodolactonization reaction to give the corresponding gamma- lactone, which is converted to prepare prepare 5-hydroxy-3,3-difluoropiperidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Difluoro-4-pentenoic acid is involved in the iodolactonization reaction to give the corresponding gamma- lactone, which is converted to prepare prepare 5-hydroxy-3,3-difluoropiperidine.

Solubility
Immiscible with water.

Notes
Incompatible with oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text