Thermo Scientific Chemicals

4-(tert-Butyl)-o-phenylenediamine, 97%, Thermo Scientific Chemicals

Catalog number: H32205.06
5 g, Each
Thermo Scientific Chemicals

4-(tert-Butyl)-o-phenylenediamine, 97%, Thermo Scientific Chemicals

Catalog number: H32205.06
5 g, Each
Quantity
Catalog number: H32205.06
also known as H32205-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
68176-57-8
IUPAC Name
4-tert-butylbenzene-1,2-diamine
Molecular Formula
C10H16N2
InChI Key
WLOSFXSXVXTKBU-UHFFFAOYSA-N
SMILES
CC(C)(C)C1=CC(N)=C(N)C=C1
Form
Powder
Melting Point (clear melt)
94.0-104.0?C
Appearance (Color)
Brown or purple
Assay (GC)
≥96.0%

Description

4-(tert-Butyl)-o-phenylenediamine, is used as a as a co-catalyst for the polymerization of o-phenylenediamine and its derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(tert-Butyl)-o-phenylenediamine, is used as a as a co-catalyst for the polymerization of o-phenylenediamine and its derivatives.

Solubility
Soluble in most common organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text