Thermo Scientific Chemicals

Dimethyl pyridine-2,6-dicarboxylate, 98%, Thermo Scientific Chemicals

Catalog number: H32251.18
50 g, Each
Thermo Scientific Chemicals

Dimethyl pyridine-2,6-dicarboxylate, 98%, Thermo Scientific Chemicals

Catalog number: H32251.18
50 g, Each
Quantity
Catalog number: H32251.18
also known as H32251-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5453-67-8
IUPAC Name
2,6-dimethyl pyridine-2,6-dicarboxylate
Molecular Formula
C9H9NO4
InChI Key
SNQQJEJPJMXYTR-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=CC(=N1)C(=O)OC
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Melting Point (clear melt)
119.5-125.5?C

Description

Eight macrocyclic bisamides and tetramides have been synthesized by reaction of dimethyl pyridine-2,6-dicarboxylate 1 with α,ω-diaminoethers in methanol as a solvent. Silver(I) coordination polymer and nine-coordinated cadmium(II) complex with dimethyl pyridine-2,6-dicarboxylate supported by solid state and electrochemical studies

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Eight macrocyclic bisamides and tetramides have been synthesized by reaction of dimethyl pyridine-2,6-dicarboxylate 1 with α,ω-diaminoethers in methanol as a solvent. Silver(I) coordination polymer and nine-coordinated cadmium(II) complex with dimethyl pyridine-2,6-dicarboxylate supported by solid state and electrochemical studies

Solubility
Soluble in water(14 g/L) (25°C). Soluble in chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text