Thermo Scientific Chemicals

1,1'-Diethyl-2,2'-carbocyanine bromide, 95%, Thermo Scientific Chemicals

Catalog number: H32253.03
1 g, Each
Thermo Scientific Chemicals

1,1'-Diethyl-2,2'-carbocyanine bromide, 95%, Thermo Scientific Chemicals

Catalog number: H32253.03
1 g, Each
Quantity
Catalog number: H32253.03
also known as H32253-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2670-67-9
IUPAC Name
1-ethyl-2-[3-(1-ethyl-1,2-dihydroquinolin-2-ylidene)prop-1-en-1-yl]quinolin-1-ium hydrobromide
Molecular Formula
C25H26BrN2
InChI Key
COPIHGSSCONEQG-UHFFFAOYSA-N
SMILES
Br.CCN1C(=CC=CC2=CC=C3C=CC=CC3=[N+]2CC)C=CC2=CC=CC=C12
Assay (special)
≥94.0 to ≤106.0% (based on elemental C, H and Br)
Elemental Analysis
Br: 17.33-19.54% (Theory: 18.44%)
Elemental Analysis
C: 65.13-73.45% (Theory: 69.29%)
Elemental Analysis
H: 5.46-6.16% (Theory: 5.81%)
Appearance (Color)
Green

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text