Thermo Scientific Chemicals

4-(Trifluoromethylsulfonyl)phenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: H32295.06
5 g, Each
Thermo Scientific Chemicals

4-(Trifluoromethylsulfonyl)phenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: H32295.06
5 g, Each
Quantity
Catalog number: H32295.06
also known as H32295-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1099597-82-6
IUPAC Name
2-(4-trifluoromethanesulfonylphenyl)acetic acid
Molecular Formula
C9H7F3O4S
InChI Key
FIEUCHICQSJBAU-UHFFFAOYSA-N
SMILES
OC(=O)CC1=CC=C(C=C1)S(=O)(=O)C(F)(F)F
Appearance (Color)
White
Assay (HPLC)
≥97.5%
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
123.0-129.0?C

Description

4-(Trifluoromethylsulfonyl)phenylacetic acid, is used as a primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethylsulfonyl)phenylacetic acid, is used as a primary and secondary intermediate.

Solubility
Slightly soluble in water, soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text