4-Chloropyridine-2-carboxylic acid, 94%, Thermo Scientific Chemicals
4-Chloropyridine-2-carboxylic acid, 94%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chloropyridine-2-carboxylic acid, 94%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Catalog number H32301.03
also known as H32301-03
Price (USD)/ Each
65.20
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
65.20
Add to cart
4-Chloropyridine-2-carboxylic acid, 94%, Thermo Scientific Chemicals
Catalog numberH32301.03
Price (USD)/ Each
65.20
-
Add to cart
Chemical Identifiers
CAS5470-22-4
IUPAC Name4-chloropyridine-2-carboxylate
Molecular FormulaC6H3ClNO2
InChI KeyNNMYRMGMVLMQAY-UHFFFAOYSA-M
SMILES[O-]C(=O)C1=CC(Cl)=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Silylated GC)≥92.0%
Appearance (Color)Pale cream to cream
FormPowder
Assay (Aqueous acid-base Titration)≥92.0 to ≤108.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only