Thermo Scientific Chemicals

Ethylamine, 2.0M in THF, Thermo Scientific Chemicals

Catalog number: H32306.AP
500 mL, Each
Thermo Scientific Chemicals

Ethylamine, 2.0M in THF, Thermo Scientific Chemicals

Catalog number: H32306.AP
500 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: H32306.AP
also known as H32306-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-04-7
IUPAC Name
ethanamine
Molecular Formula
C2H7N
InChI Key
QUSNBJAOOMFDIB-UHFFFAOYSA-N
SMILES
CCN
Appearance (Color)
Clear, colorless to pale yellow
Form
Liquid
Assay (Aqueous acid-base Titration)
1.90-2.20M

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text