Thermo Scientific Chemicals

4-Bromo-2-methylphenol, 97%, Thermo Scientific Chemicals

Catalog number: H32307.14
25 g, Each
Thermo Scientific Chemicals

4-Bromo-2-methylphenol, 97%, Thermo Scientific Chemicals

Catalog number: H32307.14
25 g, Each
Quantity
Catalog number: H32307.14
also known as H32307-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2362-12-1
IUPAC Name
4-bromo-2-methylphenol
Molecular Formula
C7H7BrO
InChI Key
IWJGMJHAIUBWKT-UHFFFAOYSA-N
SMILES
CC1=CC(Br)=CC=C1O
Form
Powder
Water Content (Karl Fischer Titration)
≤1.5%
Assay (GC)
≥96.0%
Appearance (Color)
White to pale cream
Melting Point (clear melt)
58.0-69.0?C

Description

4-Bromo-2-methylphenol, is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2-methylphenol, is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text