Thermo Scientific Chemicals

5-Bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid, 95%, Thermo Scientific Chemicals

Catalog number: H32316.03
1 g, Each
Thermo Scientific Chemicals

5-Bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid, 95%, Thermo Scientific Chemicals

Catalog number: H32316.03
1 g, Each
Quantity
Catalog number: H32316.03
also known as H32316-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
749875-16-9
IUPAC Name
5-bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid
Molecular Formula
C7H3BrF3NO2
InChI Key
XNVAVQMVAYUTBA-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC(=NC=C1Br)C(F)(F)F
Assay (Silylated GC)
≥94.0%
Appearance (Color)
Pale cream
Assay (Aqueous acid-base Titration)
≥94.0 to ≤106.0%
Melting Point (clear melt)
228.0-241.0?C
Form
Powder

Description

5-Bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water, soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text