Biphenyl-2-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Biphenyl-2-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Biphenyl-2-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H32320.06
also known as H32320-06
Price (USD)/ Each
216.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
216.00
Add to cart
Biphenyl-2-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Catalog numberH32320.06
Price (USD)/ Each
216.00
-
Add to cart
Chemical Identifiers
CAS914675-52-8
IUPAC Name2-{[1,1'-biphenyl]-2-yl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular FormulaC18H21BO2
InChI KeyWCXWQEUBHZKNMQ-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CC=CC=C1C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)79.0-85.0?C
Assay (GC)≥96.0%
Appearance (Color)Cream
Biphenyl-2-boronic acid pinacol ester, is used as a main benefits of the use of boronic esters in Suzuki-Miyaura cross-couplings- Enabling the coupling of organoboronic acids that are sensitive to hydrolytic deboronation. It includes other additional application such as a number of chiral boronic esters, such as the ones illustrated hereafter, have been used as inducers in stereoselective reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biphenyl-2-boronic acid pinacol ester, is used as a main benefits of the use of boronic esters in Suzuki-Miyaura cross-couplings- Enabling the coupling of organoboronic acids that are sensitive to hydrolytic deboronation. It includes other additional application such as a number of chiral boronic esters, such as the ones illustrated hereafter, have been used as inducers in stereoselective reactions.

Solubility
Insoluble in water. Soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
RUO – Research Use Only