Thermo Scientific Chemicals

4-Fluoro-3-nitrobenzonitrile, 97%, Thermo Scientific Chemicals

Catalog number: H32391.06
5 g, Each
Thermo Scientific Chemicals

4-Fluoro-3-nitrobenzonitrile, 97%, Thermo Scientific Chemicals

Catalog number: H32391.06
5 g, Each
Quantity
Catalog number: H32391.06
also known as H32391-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1009-35-4
IUPAC Name
4-fluoro-3-nitrobenzonitrile
Molecular Formula
C7H3FN2O2
InChI Key
LKOWKPGBAZVHOF-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC(=CC=C1F)C#N
Assay (GC)
≥96.0%
Appearance (Color)
Pale yellow to yellow
Melting Point (clear melt)
87.0-97.0?C
Form
Crystals or powder or crystalline powder

Description

4-Fluoro-3-nitrobenzonitrile, is used as a pharmaceutical intermediate. It is also used as a synthetic and fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-3-nitrobenzonitrile, is used as a pharmaceutical intermediate. It is also used as a synthetic and fine chemical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text