4-tert-Butylbenzoic anhydride, 95%, Thermo Scientific Chemicals
4-tert-Butylbenzoic anhydride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-tert-Butylbenzoic anhydride, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H32422.06
also known as H32422-06
Price (USD)/ Each
242.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
242.00
Add to cart
4-tert-Butylbenzoic anhydride, 95%, Thermo Scientific Chemicals
Catalog numberH32422.06
Price (USD)/ Each
242.00
-
Add to cart
Chemical Identifiers
CAS22201-45-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale brown
FormPowder
Assay (GC)≥94.0%
Melting Point (clear melt)75.0-84.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only