4-Bromo-3-fluorophenol, 98%, Thermo Scientific Chemicals
4-Bromo-3-fluorophenol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Bromo-3-fluorophenol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H32488.06
also known as H32488-06
Price (USD)/ Each
65.20
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
65.20
Add to cart
4-Bromo-3-fluorophenol, 98%, Thermo Scientific Chemicals
Catalog numberH32488.06
Price (USD)/ Each
65.20
-
Add to cart
Chemical Identifiers
CAS121219-03-2
IUPAC Name4-bromo-3-fluorophenol
Molecular FormulaC6H4BrFO
InChI KeyMRQYTJXVULSNIS-UHFFFAOYSA-N
SMILESOC1=CC(F)=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)69.0-75.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only