Thermo Scientific Chemicals

Sulfolane, 80% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: H32505.0F
2500 mL, Each
Thermo Scientific Chemicals

Sulfolane, 80% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: H32505.0F
2500 mL, Each
Quantity
Catalog number: H32505.0F
also known as H32505-0F
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
126-33-0
IUPAC Name
1λ⁶-thiolane-1,1-dione
Molecular Formula
C4H8O2S
InChI Key
HXJUTPCZVOIRIF-UHFFFAOYSA-N
SMILES
O=S1(=O)CCCC1
Appearance (Color)
Clear colorless to pale yellow
Water Content (Karl Fischer Titration)
18.5-21.5%
pH
8-10 (diluted to 50% aqueous)
Form
Liquid
Assay (GC)
≥99.0%

Description

Sulfolane is used in the chemical industry as a solvent for extractive distillation and chemical reactions. It is used to separate aromatic hydrocarbons (benzene, toluene and xylenes), to separate n-propyl alcohol and sec-butyl alcohol. It is also used to purify natural gas streams and for fractionation of fatty acids into saturated and unsaturated components and as a reaction solvent for the preparation of aromatic sulfonic acids, pyridines, isocyanates and pharmaceuticals. It can also be involved in the halogen exchange process and polymerization process. It is used as a plasticizer and curing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sulfolane is used in the chemical industry as a solvent for extractive distillation and chemical reactions. It is used to separate aromatic hydrocarbons (benzene, toluene and xylenes), to separate n-propyl alcohol and sec-butyl alcohol. It is also used to purify natural gas streams and for fractionation of fatty acids into saturated and unsaturated components and as a reaction solvent for the preparation of aromatic sulfonic acids, pyridines, isocyanates and pharmaceuticals. It can also be involved in the halogen exchange process and polymerization process. It is used as a plasticizer and curing agent.

Solubility
Fully miscible in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text