5-Chloro-2-hydroxybenzyl alcohol, 98%, Thermo Scientific Chemicals
5-Chloro-2-hydroxybenzyl alcohol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Chloro-2-hydroxybenzyl alcohol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
Catalog number H32533.09
also known as H32533-09
Price (USD)/ Each
207.65
Online exclusive
231.00 
Save 23.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
207.65
Online exclusive
231.00 
Save 23.35 (10%)
Add to cart
5-Chloro-2-hydroxybenzyl alcohol, 98%, Thermo Scientific Chemicals
Catalog numberH32533.09
Price (USD)/ Each
207.65
Online exclusive
231.00 
Save 23.35 (10%)
-
Add to cart
Chemical Identifiers
CAS5330-38-1
IUPAC Name4-chloro-2-(hydroxymethyl)phenol
Molecular FormulaC7H7ClO2
InChI KeyAFPSETMCMFRVPG-UHFFFAOYSA-N
SMILESOCC1=CC(Cl)=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Melting Point (clear melt)86.0-94.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only