Thermo Scientific Chemicals

4-Methyl-2-(trifluoromethyl)benzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: H32534.03
1 g, Each
Thermo Scientific Chemicals

4-Methyl-2-(trifluoromethyl)benzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: H32534.03
1 g, Each
Quantity
Catalog number: H32534.03
also known as H32534-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
120985-64-0
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (Silylated GC)
≥97.5%
Melting Point (clear melt)
136.0-142.0?C
Appearance (Color)
White to cream

Description

4-Methyl-2-(trifluoromethyl)benzoic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methyl-2-(trifluoromethyl)benzoic acid is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed co
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text