3-Methyl-1H-pyrazole-4-carboxaldehyde, 97%, Thermo Scientific Chemicals
3-Methyl-1H-pyrazole-4-carboxaldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Methyl-1H-pyrazole-4-carboxaldehyde, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number H32547.03
also known as H32547-03
Price (USD)/ Each
164.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
164.00
Add to cart
3-Methyl-1H-pyrazole-4-carboxaldehyde, 97%, Thermo Scientific Chemicals
Catalog numberH32547.03
Price (USD)/ Each
164.00
-
Add to cart
Chemical Identifiers
CAS112758-40-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Free acid (titration)≤1.5%
Melting Point (clear melt)97.0-103.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only