Thermo Scientific Chemicals

3-Vinylbenzoic acid, 96%, Thermo Scientific Chemicals

Catalog number: H32556.03
1 g, Each
Thermo Scientific Chemicals

3-Vinylbenzoic acid, 96%, Thermo Scientific Chemicals

Catalog number: H32556.03
1 g, Each
Quantity
Catalog number: H32556.03
also known as H32556-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
28447-20-3
IUPAC Name
3-ethenylbenzoic acid
Molecular Formula
C9H8O2
InChI Key
VWXZFDWVWMQRQR-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CC(C=C)=C1
Assay (Silylated GC)
>95.0%
Solution Test
(10% w/v soln. in methanol) : Clear
Appearance (Color)
White to pale yellow
Form
Powder
Assay (Aqueous acid-base Titration)
>95.0 to <105.0%

Description

Polymers were created by the imprinting of tri-O-acetyladenosine in acrylic polymer matrix with the use of methacrylic acid (MAA) or 3-vinylbenzoic acid (VBA) as a functional monomer and ethylene glycol dimethacrylate as a cross-linking agent. Nine active ester monomers based on 4-vinylbenzoic acid have been synthesized. Under free radical polymerization conditions these monomers could successfully be polymerized yielding reactive polymers

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Polymers were created by the imprinting of tri-O-acetyladenosine in acrylic polymer matrix with the use of methacrylic acid (MAA) or 3-vinylbenzoic acid (VBA) as a functional monomer and ethylene glycol dimethacrylate as a cross-linking agent. Nine active ester monomers based on 4-vinylbenzoic acid have been synthesized. Under free radical polymerization conditions these monomers could successfully be polymerized yielding reactive polymers

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat, light, ultraviolet radiations, free radical initiators.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text