Thermo Scientific Chemicals

Agarose Beads 6% B-CL (Cross-linked)

Catalog number: H32568.AU
1000 mL, Each
Thermo Scientific Chemicals

Agarose Beads 6% B-CL (Cross-linked)

Catalog number: H32568.AU
1000 mL, Each
Quantity
Catalog number: H32568.AU
also known as H32568-AU
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
62610-50-8
Appearance (Color)
White to pale cream
Form
Spherical beads
Assay (unspecified)
Agarose content: 6%
Comment
Antimicrobial agent: 20% ethanol
Comment
Stable working pH range: 3-13

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text